Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Desmuslin Antibody (OTI7H6), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271736A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271736A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271736A48 Supplier Novus Biologicals Supplier No. NBP271736AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha Desmuslin Monoclonal specifically detects alpha Desmuslin in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen alpha Desmuslin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7H6
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias desmuslin, DMNEC 1.4.99.1, EC 2.6.1.16, KIAA0353synemin beta, SYNDesmuslin, synemin, synemin alpha, synemin, intermediate filament protein
Gene Symbols SYNM
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 329-580 of human alpha Desmuslin (NP_663780) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23336.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.