Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Dystroglycan Antibody (2238), Novus Biologicals™
SDP

Catalog No. NBP149634 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149634 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149634 Supplier Novus Biologicals Supplier No. NBP149634
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

Alpha Dystroglycan Monoclonal specifically detects Alpha Dystroglycan in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoassay.

Specifications

Antigen Alpha Dystroglycan
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2238
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay
Gene Accession No. O18738
Gene Alias A3a, AGRNR, DAG156DAG, dystroglycan, dystroglycan 1 (dystrophin-associated glycoprotein 1), Dystrophin-associated glycoprotein 1
Gene Symbols DAG1
Host Species Mouse
Immunogen Monoclonal antibody 2238 recognizes a glycoepitope unique to brain alpha-dystroglycan.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1605
Test Specificity The monoclonal antibody 2238 is specific for a glycoepitope on brain bovine alpha-dystroglycan, which is absent on alpha-dystroglycan expressed in all other tissues.
Target Species Human, Mouse, Rat, Bovine, Rabbit
Content And Storage Store at 4°C.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.