Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP Antibody (AF5), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB030139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB030139 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB030139 Supplier Novus Biologicals Supplier No. NBP282221C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen alpha-Fetoprotein/AFP
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone AF5
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Host Species Rabbit
Immunogen AF5 was prepared by immunizing female BALB/c mice with human amniotic fluid containing alpha-Fetoprotein/AFP, from a pregnancy affected by a neural tube defect, diluted and emulsified in Freund's complete adjuvant.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 174
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.