Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP Antibody (AFP-11), DyLight 405
SDP

Catalog No. NB500524V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500524V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500524V Supplier Novus Biologicals Supplier No. NB500524V

Mouse Monoclonal Antibody

alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen alpha-Fetoprotein/AFP
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AFP-11
Conjugate DyLight 405
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Gene Symbols AFP
Host Species Mouse
Immunogen purified alpha-Fetoprotein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cancer Stem Cells, GPCR, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 174
Test Specificity This antibody (clone AFP-11) reacts with human alpha-Fetoprotein (AFP), a 70 kDa oncofetal antigen. AFP is a major fetal plasma protein, but is not present in healthy adult tissues. Elevated AFP concentrations in adult plasma may be an early marker of hepatocellular carcinoma or teratoblastoma, while high concentrations in amniotic fluid may indicate severe congenital defects of a fetus.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.