Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP Antibody (AFP/7007R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB187245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187245 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187245 Supplier Novus Biologicals Supplier No. NBP314053JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen alpha-Fetoprotein/AFP
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AFP/7007R
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Host Species Rabbit
Immunogen Recombinant human full-length alpha-Fetoprotein/AFP protein (Uniprot: P02771 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 174
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.