Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP Antibody (C3), PerCP, Novus Biologicals™
SDP

Catalog No. NBP234577P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234577P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234577P Supplier Novus Biologicals Supplier No. NBP234577PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human, Porcine, Canine, Monkey, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Flow (Intracellular), Knockout Validated.

Specifications

Antigen alpha-Fetoprotein/AFP
Applications Flow Cytometry, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C3
Conjugate PerCP
Dilution Flow Cytometry, Flow (Intracellular), Knockout Validated
Gene Alias Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Gene Symbols AFP
Host Species Mouse
Immunogen Alpha feto protein (AFP) Purified from serum of a hepatoma patient (Uniprot: P02771)
Molecular Weight of Antigen 70 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 174
Test Specificity It recognizes an oncofetal glycoprotein with a single chain of 70kDa, which is identified as alpha-fetoprotein (AFP) (ISOBM TD-2 workshop). This monoclonal antibody is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.
Target Species Human, Pig, Canine, Primate, Bovine (Negative), Canine (Negative), Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.