Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP Antibody (CL9772), Novus Biologicals™
SDP

Catalog No. NB238233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB238233 100 μg
NB238018 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB238233 Supplier Novus Biologicals Supplier No. NBP321160100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen alpha-Fetoprotein/AFP
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL9772
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Host Species Mouse
Immunogen This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of P02771. Actual immunogen sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 174
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.