Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP Antibody (OTI2A9), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP270103C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270103C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270103C Supplier Novus Biologicals Supplier No. NBP270103C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen alpha-Fetoprotein/AFP
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2A9
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Gene Symbols AFP
Host Species Mouse
Immunogen Full length recombinant protein of human alpha-Fetoprotein/AFP (NP_001125) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 174.0
Target Species Human, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.