Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ alpha-Fetoprotein/AFP Antibody Pair [HRP]
SDP

Catalog No. p-7183129 Shop All R&D Systems Products
Click to view available options
Quantity:
15 Plates
5 Plates

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human alpha-Fetoprotein/AFP.

Specifications

Assay Range 11 pg/mL
Assay Sensitivity 10.94-700 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human alpha-Fetoprotein/AFP.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Gene ID (Entrez) 174
Gene Symbol AFP
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-alpha-Fetoprotein/AFP monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.1 mg/mL of mouse anti-alpha-Fetoprotein/AFP monoclonal antibody.
  • Standard: Each vial contains 38 ng of recombinant alpha-Fetoprotein/AFP.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
    Storage Requirements Storage is content dependent.
    Target alpha-Fetoprotein/AFP
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.7 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.