Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A2BP1 Antibody (D8H8), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$207.00 - $501.50
Specifications
| Antigen | A2BP1 |
|---|---|
| Clone | D8H8 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:20, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 30001398) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP213169SS
![]() |
Novus Biologicals
NBP213169SS |
0.025 mL |
Each for $207.00
|
|
|||||
NBP213169
![]() |
Novus Biologicals
NBP213169 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
A2BP1 Monoclonal specifically detects A2BP1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| A2BP1 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Epigenetics, Neuroscience | |
| Q9NWB1, Q9JJ43 | |
| 54715 | |
| Mouse recombinant A2BP1 [Swiss-Prot# Q9JJ43] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| D8H8 | |
| Western Blot 1:1000, Simple Western 1:20, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 30001398) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 2BP1, A2BP, A2BP1 Hexaribonucleotide-binding protein 1, Ataxin-2-binding protein 1, FOX-1, FOX1 hexaribonucleotide binding protein 1, Fox-1 homolog A, HRNBP1 ataxin 2-binding protein 1, RNA binding protein fox-1 homolog 1, RNA binding protein, fox-1 homolog (C. elegans) 1 | |
| RBFOX1 | |
| IgG1 κ | |
| Protein G purified | |
| Does not cross react with paralogue FOX2 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title