Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Fodrin Antibody (SPTAN1/3352) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119074 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119074 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119074 Supplier Novus Biologicals Supplier No. NBP308348
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alpha Fodrin Monoclonal specifically detects Alpha Fodrin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Alpha Fodrin
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone SPTAN1/3352
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2
Host Species Mouse
Immunogen Recombinant fragment of human Alpha Fodrin protein (around aa 2351-2475) (exact sequence is proprietary))
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6709
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.