Learn More
alpha-Galactosidase A/GLA Antibody (001), Alexa Fluor™ 700, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | alpha-Galactosidase A/GLA |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1 |
| Conjugate | Alexa Fluor 700 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Agalsidase, agalsidase alfa, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, alpha-D-galactoside galactohydrolase 1, alpha-gal A, alpha-galactosidase A, EC 3.2.1, EC 3.2.1.22, GALA, galactosidase, alpha, melibiase |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human alpha-Galactosidase A/GLA (AAP36507.1; Met 1-Leu 429). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.