Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Galactosidase A/GLA Antibody (201), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB037065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037065 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037065 Supplier Novus Biologicals Supplier No. NBP290048AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

alpha-Galactosidase A/GLA Monoclonal specifically detects alpha-Galactosidase A/GLA in Human samples. It is validated for Western Blot.

Specifications

Antigen alpha-Galactosidase A/GLA
Applications Western Blot
Classification Monoclonal
Clone 201
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Agalsidase, agalsidase alfa, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, alpha-D-galactoside galactohydrolase 1, alpha-gal A, alpha-galactosidase A, EC 3.2.1, EC 3.2.1.22, GALA, galactosidase, alpha, melibiase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human alpha-Galactosidase A/GLA (AAP36507.1; Met1-Leu429).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2717
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.