Learn More
Invitrogen™ alpha Galactosidase Monoclonal Antibody (2H7G10)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human alpha-Galactosidase A/GLA.
Specifications
Specifications
| Antigen | alpha Galactosidase |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2H7G10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | GLA |
| Gene Accession No. | P06280 |
| Gene Alias | Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; Melibiase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.