Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha GalCer:CD1d Complex Monoclonal Antibody (L363), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$141.00 - $405.00

Specifications

Antigen alpha GalCer:CD1d Complex
Clone L363
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-107-14
Encompass_Preferred
View Documents
Invitrogen™
12201980
25 μg
Each for $141.00
Only null left
Add to Cart
 
50-107-15
Encompass_Preferred
View Documents
Invitrogen™
12201982
100 μg
Each for $405.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: This L363 monoclonal antibody reacts with alpha-galactosylceramide bound to the mouse MHC class I-like protein CD1d. This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Valpha14Jalpha18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation. The L363 antibody has been reported to neutralize iNKT cell activation by alpha-galactosylceramide:CD1d complexes. Applications Reported: This L363 antibody has been reported for use in flow cytometric analysis. Applications Tested: This L363 antibody has been tested by flow cytometric analysis of alpha-galactosylceramide-loaded mouse CD1d-transfected cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Va14Ja18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation.
Specifications

Specifications

alpha GalCer:CD1d Complex
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
3'-phosphoadenosine-5'-phosphosulfate:Galcer sulfotransferase; 3'-phosphoadenylylsulfate:galactosylceramide 3'-sulfotransferase; 3'-phosphoadenylylsulfate-galactosylceramide 3'-sulfotransferase; AI747460; Antigen-presenting glycoprotein CD1d1; CD1.1; Cd1a; Cd1d; CD1d.1; Cd1d1; CD1d1 antigen; cerebroside sulfotransferase; Cst; Gal3st1; galactose-3-O-sulfotransferase 1; galactosylceramide sulfotransferase; GalCer sulfotransferase; Gcst; Ly-38; T-cell surface glycoprotein CD1d1
Cd1d1, Gal3st1
Primary
4°C, store in dark, DO NOT FREEZE!
Cd1d1
L363
Flow Cytometry
PE
Mouse
Mouse
P11609, Q9JHE4
12479, 53897
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.