Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Internexin Antibody (1D2), Novus Biologicals™
SDP

Catalog No. NB018149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018149 0.125 mL
NB300216 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018149 Supplier Novus Biologicals Supplier No. NB3002160.125ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-Internexin Monoclonal specifically detects alpha-Internexin in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.

Specifications

Antigen alpha-Internexin
Applications Immunohistochemistry (Free Floating), Immunocytochemistry, Immunohistochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1D2
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Immunohistochemistry Free-Floating 1:5000
Formulation 50% PBS, 50% Glycerol with 5mM Sodium Azide
Gene Alias 66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1
Gene Symbols INA
Host Species Mouse
Immunogen Purified recombinant rat alpha-internexin expressed in and purified from E. coli.
Molecular Weight of Antigen 64-66 kDa
Purification Method Affinity purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9118
Test Specificity Human and all mammalian species tested to date.
Target Species Human, Feline, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.