Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Internexin Antibody (1D2), Novus Biologicals™

Mouse Monoclonal Antibody
$149.33 - $480.74
Specifications
| Antigen | alpha-Internexin |
|---|---|
| Clone | 1D2 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Immunohistochemistry Free-Floating 1:5000 |
| Applications | Immunohistochemistry (Free Floating), Immunocytochemistry, Immunohistochemistry, Immunofluorescence, Western Blot |
Description
alpha-Internexin Monoclonal specifically detects alpha-Internexin in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Specifications
| alpha-Internexin | |
| 1 mg/ml | |
| Immunohistochemistry (Free Floating), Immunocytochemistry, Immunohistochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| 66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1 | |
| INA | |
| IgG1 | |
| Affinity purified | |
| Human and all mammalian species tested to date. |
| 1D2 | |
| Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Immunohistochemistry Free-Floating 1:5000 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Human, Feline, Mouse, Rat | |
| 9118 | |
| Purified recombinant rat alpha-internexin expressed in and purified from E. coli. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 64-66 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title