Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Internexin Antibody, Novus Biologicals™
SDP

Catalog No. NB018358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018358 0.025 mL
NBP105208 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018358 Supplier Novus Biologicals Supplier No. NBP1052080.025ML
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

alpha-Internexin Polyclonal specifically detects alpha-Internexin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.

Specifications

Antigen alpha-Internexin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating), Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000
Formulation PBS with 5mM Sodium Azide
Gene Alias 66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1
Gene Symbols INA
Host Species Chicken
Immunogen Purified full length recombinant rat alpha-internexin expressed in and purified from E. coli.
Molecular Weight of Antigen 65 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9118
Target Species Pig, Equine, Bovine, Human, Rat, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.