Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ alpha-Mangostin

Catalog No. 685610
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits proliferation and induces apoptosis selectively in pancreatic CSCs and cancer cell lines

α-Mangostin is a xanthone derivative. Inhibits proliferation and induces apoptosis selectively in pancreatic CSCs and cancer cell lines. Also suppresses the invasive and metastatic ability of pancreatic CSCs by modulation of cadherin expression and suppressing the transcription factors Snail and Slug. Attenuates the expression of Nanog and pluripotency promoting transcription factors, and inhibits Nanog binding. Downregulates the PI 3-K/Akt pathway. Inhibits self-renewal of CSCs isolated from human primary tumors and KrasG12D mice. Inhibits MRSA and VRE. Antibacterial, antifungal, antimalarial, anticarcinogenic, antiatherogenic, anti-inflammatory, antioxidant and neuroprotective.

Specifications

Chemical Name or Material 1,3,6-Trihydroxy-7-methoxy-2,8-bis(3-methyl-2-buten-1-yl)-9H-xanthen-9-one
CAS 1551491
Quantity 10 mg
Molecular Formula C24H26O6
Formula Weight 410.47
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.