Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ alpha-Mangostin
Click to view available options
Quantity:
10 mg
Description
α-Mangostin is a xanthone derivative. Inhibits proliferation and induces apoptosis selectively in pancreatic CSCs and cancer cell lines. Also suppresses the invasive and metastatic ability of pancreatic CSCs by modulation of cadherin expression and suppressing the transcription factors Snail and Slug. Attenuates the expression of Nanog and pluripotency promoting transcription factors, and inhibits Nanog binding. Downregulates the PI 3-K/Akt pathway. Inhibits self-renewal of CSCs isolated from human primary tumors and KrasG12D mice. Inhibits MRSA and VRE. Antibacterial, antifungal, antimalarial, anticarcinogenic, antiatherogenic, anti-inflammatory, antioxidant and neuroprotective.
Specifications
Specifications
Chemical Name or Material | 1,3,6-Trihydroxy-7-methoxy-2,8-bis(3-methyl-2-buten-1-yl)-9H-xanthen-9-one |
CAS | 1551491 |
Quantity | 10 mg |
Molecular Formula | C24H26O6 |
Formula Weight | 410.47 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction