Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Smooth Muscle Actin Antibody (132D3), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB161123 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161123 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB161123 Supplier Novus Biologicals Supplier No. NBP259429JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-Smooth Muscle Actin Monoclonal antibody specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen alpha-Smooth Muscle Actin
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 132D3
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
Host Species Mouse
Immunogen Alpha-Smooth Muscle Actin Antibody (132D3) is made to a recombinant human ACTA2 (3-377aa) purified from E. coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 59
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.