Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Smooth Muscle Actin Antibody (1A4), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB304954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304954 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304954 Supplier Novus Biologicals Supplier No. FAB1420K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat samples. It is validated for CyTOF, Dual RNAScope ISH-IHC, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry, Western Blot.

Specifications

Antigen alpha-Smooth Muscle Actin
Applications CyTOF, Dual RNAScope ISH-IHC, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1A4
Conjugate DyLight 488
Dilution Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC
Formulation 50mM Sodium Borate
Gene Alias AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
Host Species Mouse
Immunogen Peptide corresponding to residues EEEDSTALVC of the N-term of human alpha-smooth muscle actin (1)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 59
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.