Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Smooth Muscle Actin Antibody (BLR082G), Novus Biologicals™
SDP

Catalog No. NB002998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002998 Supplier Novus Biologicals Supplier No. NBP278836
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.

Specifications

Antigen alpha-Smooth Muscle Actin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone BLR082G
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry 1:100 - 1:500
Formulation Borate Buffered Saline (BBS) pH 8.2 with 0.09% Sodium Azide
Gene Alias 1a4 sma, 1a4 smooth muscle actin, AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha sma 1a4, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, anti-alpha SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, sma 1a4, smooth muscle actin
Gene Symbols ACTA2
Host Species Rabbit
Immunogen Alpha-Smooth Muscle Actin Antibody (BLR082G) is a recombinant monoclonal made to a peptide representing a region between residue between 1 and 50 of human aortic smooth muscle actin using the numbering given in entry NP_001135417.1 (Gene ID 59).
Molecular Weight of Antigen 42 kDa
Purification Method Tissue culture supernatant
Quantity 100 μL
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 59
Target Species Human, Mouse
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.