Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Smooth Muscle Actin Antibody, Novus Biologicals™
SDP

Catalog No. NB6001434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001434 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001434 Supplier Novus Biologicals Supplier No. NB6001434

Rabbit Polyclonal Antibody has been used in 4 publications

alpha-Smooth Muscle Actin Polyclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Bovine, Chicken, Feline, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen alpha-Smooth Muscle Actin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:25, Immunohistochemistry-Paraffin 1:25, Immunohistochemistry-Frozen
Formulation PBS (pH 7.6) and 0.2% BSA with 15mM Sodium Azide
Gene Accession No. P62736
Gene Alias AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
Gene Symbols ACTA2
Host Species Rabbit
Immunogen Alpha-Smooth Muscle Actin Antibody is made to a synthetic peptide from N-terminus of human Alpha-smooth muscle isoform of actin.
Molecular Weight of Antigen 42 kDa
Purification Method Affinity Purified
Quantity 500 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 59
Test Specificity Cross-reacts with Rabbit, Human, Mouse, Rat, Cow, Baboon and Chicken.
Target Species Human, Mouse, Rat, Bovine, Chicken, Feline, Primate, Rabbit, Sheep
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.