Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Smooth Muscle Actin Mouse, Clone: SPM332, Novus Biologicals™
SDP

Catalog No. NBP232808F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232808F 0.02 mg
NBP232808A 0.1 mg
NBP232808B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP232808F Supplier Novus Biologicals Supplier No. NBP2328080.02MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Baboon, Chimpanzee, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen alpha-Smooth Muscle Actin
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM332
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P62736
Gene Alias AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
Gene Symbols ACTA2
Host Species Mouse
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Molecular Weight of Antigen 42 kDa
Purification Method Protein A purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 59
Test Specificity Actin is a major component of the cytoskeleton and is present in most cell types. This MAb is highly specific to actin from smooth muscles. Its epitope lies in the first four N-terminal amino acids. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actin and anti-smooth muscle actin and are negative with anti-myogenin.
Target Species Human, Mouse, Rat, Pig, Baboon, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Primate, Rabbit, Sheep
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.