Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Synuclein Antibody (017), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB037085 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037085 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037085 Supplier Novus Biologicals Supplier No. NBP290051AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen alpha-Synuclein
Applications Western Blot, ELISA
Classification Monoclonal
Clone 17
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Host Species Rabbit
Immunogen This alpha-Synuclein Antibody (017) was developed against rabbit immunized with purified, recombinant human alpha-Synuclein (P37840-1; Met1-Ala140).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6622
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.