Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Synuclein Antibody (2A7), Novus Biologicals™
SDP

Catalog No. NB018353 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018353 0.025 mL
NBP105194 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018353 Supplier Novus Biologicals Supplier No. NBP1051940.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.

Specifications

Antigen alpha-Synuclein
Applications Immunohistochemistry (Free Floating), Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2A7
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000
Formulation 50% PBS, 50% Glycerol with 5mM Sodium Azide
Gene Accession No. P37840
Gene Alias Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Gene Symbols SNCA
Host Species Mouse
Immunogen Full length human recombinant protein with the epitope from amino acids 61-95
Molecular Weight of Antigen 15 kDa
Purification Method Immunogen affinity purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6622
Target Species Pig, Human, Bovine, Rat, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.