Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Synuclein Antibody (2A7), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$244.50 - $584.50
Specifications
Antigen | alpha-Synuclein |
---|---|
Clone | 2A7 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:1000, Simple Western 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000 |
Applications | Immunohistochemistry (Free Floating), Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry |
Description
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Specifications
alpha-Synuclein | |
1 mg/ml | |
Immunohistochemistry (Free Floating), Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Pig, Human, Bovine, Rat, Mouse | |
Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
SNCA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
2A7 | |
Western Blot 1:1000, Simple Western 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
P37840 | |
6622 | |
IgG1 | |
Immunogen affinity purified | |
15 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title