Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Synuclein Antibody (OTI2F6), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP271293A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271293A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271293A30 Supplier Novus Biologicals Supplier No. NBP271293AF350

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen alpha-Synuclein
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2F6
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Gene Symbols SNCA
Host Species Mouse
Immunogen This alpha-Synuclein Antibody (OTI2F6) was developed against full length human recombinant protein of human alpha-Synuclein(NP_000336) produced in HEK293T cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6622.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.