Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-synuclein N103 fragment Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | alpha-synuclein N103 fragment |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN2260-25UG
|
MilliporeSigma
ABN226025UG |
25 μg |
Each for $192.35
|
|
|||||
ABN2260-100
|
MilliporeSigma
ABN2260100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-alpha-synuclein N103 fragment Antibody, Cat. No. ABN2260, is a highly specific rabbit polyclonal antibody that targets alpha-synuclein and has been tested for use in Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.Specifications
alpha-synuclein N103 fragment | |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
Purified rabbit polyclonal antibody in PBS with 0.02 mg/ml sodium azide and 40% glycerol. | |
NP_000336 | |
KLH-conjugated linear peptide corresponding to nine amino acids that precede the asparagine endopeptidase (AEP)-cleavage site at N103. | |
Primary | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Non-A beta component of AD amyloid;Non-A4 component of amyloid precursor;NACP | |
SNCA;NACP;PARK1 | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody detects human alpha-synuclein N103 fragment. | |
∽38 kD observed (GFP Fusion protein); 14.46 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title