Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha TLR10 Mouse anti-Human, Unconjugated, Clone: 5C2C5, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | alpha TLR10 |
---|---|
Clone | 5C2C5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF20942MI
|
MilliporeSigma
MABF209425UG |
25 μg |
Each for $189.29
|
|
|||||
MABF20941MI
|
MilliporeSigma
MABF2094100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
alpha TLR10 Monoclonal antibody specifically detects alpha TLR10 in Human samples. It is validated for Flow Cytometry, Inhibits Activity/Function.Specifications
alpha TLR10 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
CD290 | |
Recombinant fragment corresponding to 455 amino acids from the N-terminal, extracellular domain of human Toll-like receptor 10. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
94.56 kDa calculated. |
5C2C5 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_001017388 | |
TLR10, UNQ315/PRO358 | |
IgG1 κ | |
Protein G Purified | |
Clone 5C2C5 is a mouse monoclonal antibody that detects human Toll-like receptor 10. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title