Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin 3c Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB342742 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342742 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342742 Supplier Novus Biologicals Supplier No. NBP281701MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

alpha Tubulin 3c Polyclonal antibody specifically detects alpha Tubulin 3c in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen alpha Tubulin 3c
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Alpha-tubulin 2, Alpha-tubulin 3C/D, bA408E5.3, TUBA2Tubulin alpha-2 chain, TUBA3D, tubulin alpha-3C/D chain, tubulin, alpha 2, tubulin, alpha 3c
Host Species Rabbit
Immunogen Rabbit polyclonal alpha Tubulin 3c antibody was raised against a 16 amino acid peptide near the amino terminus of human alpha Tubulin 3c. The immunogen is located within amino acids 140 - 190 of alpha Tubulin 3c. Amino Acid Squence: MERLSVDYGKKSKLEF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7278
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.