Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin 4a Antibody (B-5-1-2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1201304A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1201304A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1201304A30 Supplier Novus Biologicals Supplier No. NB12011304AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha Tubulin 4a Monoclonal specifically detects alpha Tubulin 4a in Human, Mouse, Rat, Bovine, Chicken, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen alpha Tubulin 4a
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone B-5-1-2
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias alpha 1 (testis specific), Alpha-tubulin 1, FLJ30169, H2-ALPHA, Testis-specific alpha-tubulin, TUBA1, Tubulin alpha-1 chain, tubulin alpha-4A chain, Tubulin H2-alpha, tubulin, alpha 1, tubulin, alpha 4a
Gene Symbols TUBA4A
Host Species Mouse
Immunogen Sarkosyl-resistant filaments from Stronglycentrotus purpuratus (sea urchin).
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7277
Test Specificity Recognizes an epitope located at the C-terminal end of the alpha-tubulin isoform in a variety of organisms.
Target Species Human, Mouse, Rat, Bovine, Chicken, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.