Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin 4a Antibody, Novus Biologicals™
SDP

Catalog No. NBP221604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221604 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221604 Supplier Novus Biologicals Supplier No. NBP221604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

alpha Tubulin 4a Polyclonal antibody specifically detects alpha Tubulin 4a in Human, Mouse, Rat, Chicken, Drosophila, Eukaryote samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen alpha Tubulin 4a
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
Gene Alias alpha 1 (testis specific), Alpha-tubulin 1, FLJ30169, H2-ALPHA, Testis-specific alpha-tubulin, TUBA1, Tubulin alpha-1 chain, tubulin alpha-4A chain, Tubulin H2-alpha, tubulin, alpha 1, tubulin, alpha 4a
Gene Symbols TUBA4A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human alpha Tubulin 4a. The exact sequence is proprietary.
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7277
Test Specificity Human, Mouse, Rat, Drosophila, Chicken. Expected to cross react based on sequence identity: Bovine (100%), Xenopus tropicalis (100%), Sheep (100%).
Target Species Human, Mouse, Rat, Chicken, Drosophila
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.