Learn More
Description
Specifications
Specifications
| Antigen | alpha Tubulin |
| Applications | CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | DM1A |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000, Simple Western 1:50, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50000-1:100000, Immunoprecipitation 1:50-1:100, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, Immunomicroscopy, Flow (Intracellular), CyTOF-ready |
| Gene Alias | alpha tubulin loading control, alpha tubulin monoclonal, Alpha-tubulin 3, B-ALPHA-1, DM1A, DM1A anti-tubulin, dm1a monoclonal, DM1a tubulin, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA1 monoclonal, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin monoclonal, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific |
| Gene Symbols | TUBA1A |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
