Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin Antibody (DM1A), PE, Novus Biologicals™
SDP

Catalog No. NB100690PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100690PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100690PE Supplier Novus Biologicals Supplier No. NB100690PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha Tubulin Monoclonal specifically detects alpha Tubulin in Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Fungi, Goat, Guinea Pig, Hamster, Human, Monkey, Mouse, Parasite, Pig, Primate, Rabbit, Rat, Xenopus species, Yeast samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot.

Specifications

Antigen alpha Tubulin
Applications CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DM1A
Conjugate PE
Dilution Flow Cytometry
Gene Alias Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Gene Symbols TUBA1A
Host Species Mouse
Immunogen This alpha Tubulin Antibody (DM1A) was developed against native chicken brain microtubules.
Molecular Weight of Antigen 50 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7846
Test Specificity This alpha Tubulin Antibody (DM1A) does not cross-react with beta Tubulin.
Target Species Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Fungi, Goat, Guinea Pig, Hamster, Human, Monkey, Mouse, Parasite, Pig, Primate, Rabbit, Rat, Xenopus species, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.