Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin Antibody, Novus Biologicals™
SDP

Catalog No. NB287579 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287579 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287579 Supplier Novus Biologicals Supplier No. NBP323471
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen alpha Tubulin
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 -1:2000, Immunocytochemistry/Immunofluorescence : 1:100 - 1:200
Formulation 50% Glycerol/PBS, 1% BSA
Gene Alias Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Host Species Rabbit
Immunogen A peptide corresponding to dC2-alpha Tubulin (Uniprot#: Q71U36 )
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7846
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.