Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin Antibody (SR1897), Novus Biologicals™
SDP

Catalog No. NB258105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258105 50 μg
NB258205 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258105 Supplier Novus Biologicals Supplier No. NBP32267150UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in Yeast samples. It is validated for Western Blot

Specifications

Antigen alpha Tubulin
Applications Western Blot
Classification Monoclonal
Clone SR1897
Conjugate Unconjugated
Dilution Western Blot 1:3000-1:10000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Host Species Rabbit
Immunogen A synthesized peptide derived from yeast alpha Tubulin (Uniprot #: P09733)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7846
Target Species Yeast
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.