Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin Antibody (TU-01), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB500333AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500333AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500333AF Supplier Novus Biologicals Supplier No. NB500333AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Porcine, Mammal, Plant, Turkey, Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Tubulin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TU-01
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Gene Symbols TUBA1A
Host Species Mouse
Immunogen This alpha Tubulin Antibody (TU-01) was developed against a fraction of tubulin purified from pig brain by two cycles of polymerization-depolymerization.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7846
Test Specificity This alpha Tubulin Antibody (TU-01) recognizes the defined epitope (aa 65-97) on N-terminal structureal domain of alpha tubulin.
Target Species Human, Mouse, Rat, Pig, Mammalia, Plant, Turkey, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.