Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin Antibody (YL1/2), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB600506Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600506Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600506Z Supplier Novus Biologicals Supplier No. NB600506AF700
Add to Cart
Add to Cart

Rat Monoclonal Antibody

alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Avian, C. elegans, Drosophila, Invertebrate, Mammal, Primate, Monkey, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen alpha Tubulin
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone YL1/2
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Gene Symbols TUBA1A
Host Species Rat
Immunogen This alpha Tubulin Antibody (YL1/2) was developed against full length native protein (purified) (S. cerevisiae).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7846
Test Specificity Human, mouse, rat, monkey, S. cerevisiae, S. pombe, Slime moulds, Allium and birds. Expected to react with most eukaryotes due to sequence identity. Drosophila reactivity reported in scientific literature (PMID: 24019759) Other species have not been tested.
Target Species Human, Mouse, Rat, Avian, C. elegans, Drosophila, Invertebrate, Mammalia, Primate, Monkey, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.