Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin Antibody (YOL1/34) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1001639SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001639SS 0.05 mg
NB1001639 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001639SS Supplier Novus Biologicals Supplier No. NB1001639SS
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 18 publications

alpha Tubulin Monoclonal specifically detects alpha Tubulin in Drosophila, Fungi, Human, Mouse, Rat, Yeast samples. It is validated for ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Radioimmune Assays (RIA), Western Blot.

Specifications

Antigen alpha Tubulin
Applications ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone YOL1/34
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunoassay 1:100-1:2000, Flow (Intracellular)
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Accession No. Q71U36
Gene Alias alpha tubulin loading control, alpha tubulin monoclonal, Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA1 monoclonal, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin monoclonal, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Gene Symbols TUBA1A
Host Species Rat
Immunogen This alpha Tubulin Antibody (YOL1/34) was made from yeast tubulin.
Molecular Weight of Antigen 50 kDa
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Biology, Loading Controls, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7846
Test Specificity This alpha Tubulin Antibody (YOL1/34) detects an epitope on alpha tubulin between the amino acids 414-422. It has a higher affinity for fixed microtubules (formaldehyde or glutaraldehyde) than native microtubules when performing IHC.
Target Species Drosophila, Mouse, Fungi, Human, Mouse, Yeast, Rat, Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.