Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin Antibody (YOL1/34) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 18 publications
$219.50 - $536.00
Specifications
Antigen | alpha Tubulin |
---|---|
Clone | YOL1/34 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunoassay 1:100-1:2000, Flow (Intracellular) |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1001639SS
![]() |
Novus Biologicals
NB1001639SS |
0.05 mg |
Each for $219.50
|
|
|||||
NB1001639
![]() |
Novus Biologicals
NB1001639 |
0.25 mg |
Each for $536.00
|
|
|||||
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Drosophila, Fungi, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Radioimmunoassay, Flow (Intracellular).Specifications
alpha Tubulin | |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rat | |
Cell Biology, Loading Controls, Phospho Specific, Stem Cell Markers | |
Q71U36 | |
7846 | |
This alpha Tubulin Antibody (YOL1/34) was made from yeast tubulin. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
50 kDa |
YOL1/34 | |
Western Blot 1:100-1:2000, ELISA 10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunoassay 1:100-1:2000, Flow (Intracellular) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Drosophila, Fungi, Yeast | |
alpha tubulin loading control, alpha tubulin monoclonal, Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA1 monoclonal, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin monoclonal, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
TUBA1A | |
IgG2a | |
Protein G purified | |
This alpha Tubulin Antibody (YOL1/34) detects an epitope on alpha tubulin between the amino acids 414-422. It has a higher affinity for fixed microtubules (formaldehyde or glutaraldehyde) than native microtubules when performing IHC. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title