Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaA Crystallin/CRYAA Antibody (1H3.B8), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP212875C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212875C 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212875C Supplier Novus Biologicals Supplier No. NBP212875C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AlphaA Crystallin/CRYAA Monoclonal specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AlphaA Crystallin/CRYAA
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1H3.B8
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4
Gene Symbols CRYAA
Host Species Mouse
Immunogen Native Alpha Crystallin
Purification Method Protein G purified
Quantity 200 μL
Research Discipline Membrane Trafficking and Chaperones, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1409
Test Specificity Detects approx 20kDa. Does not cross-react with aB-crystallin, Beta L-crystallin, BH- crystallin, gamma-crystallin, HSP25, HSP27 or HSP47 proteins.
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.