Learn More
AlphaA Crystallin/CRYAA Antibody (c9F2) - BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | AlphaA Crystallin/CRYAA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | c9F2 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, ELISA 1:100-1:2000 |
| Gene Accession No. | P02489 |
| Gene Alias | CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4 |
| Gene Symbols | CRYAA |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.