Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AlphaA Crystallin/CRYAA Antibody (c9F2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$304.50 - $706.50
Specifications
| Antigen | AlphaA Crystallin/CRYAA |
|---|---|
| Clone | c9F2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, ELISA 1:100-1:2000 |
| Applications | ELISA, Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018073
![]() |
Novus Biologicals
NB120148210.025ML |
0.025 mL |
Each for $304.50
|
|
|||||
NB120148215
![]() |
Novus Biologicals
NB120148210.05ML |
0.05 mL |
Each for $426.00
|
|
|||||
NB12014821
![]() |
Novus Biologicals
NB12014821 |
0.1 mL |
Each for $706.50
|
|
|||||
Description
AlphaA Crystallin/CRYAA Monoclonal specifically detects AlphaA Crystallin/CRYAA in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
| AlphaA Crystallin/CRYAA | |
| 1.0 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Human, Rat | |
| CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4 | |
| CRYAA | |
| IgG1 κ | |
| NB120-14821 recognizes alpha A crystallin, a 20kDa water-soluble major structural protein and member of the heat shock protein family, designated HSPB4, which acts as a molecular chaperone and is believed to play a role in the maintenance of ocular lens transparency/refractive index. Besides the involvement of both alpha A and alpha B crystallin in the prevention of protein precipitation and cataract formation in the eye, alpha crystallins have also been demonstrated in other tissues including the heart, brain, kidney, spleen and thymus. |
| c9F2 | |
| Western Blot 1:1000-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| P02489 | |
| 1409 | |
| E. coli-derived recombinant protein corresponding to full-length (aa1-173) human crystallin alpha A. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title