Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaA Crystallin/CRYAA Antibody (OTI3B12), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP270450IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270450IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270450IR Supplier Novus Biologicals Supplier No. NBP270450IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AlphaA Crystallin/CRYAA Monoclonal specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AlphaA Crystallin/CRYAA
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3B12
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4
Gene Symbols CRYAA
Host Species Mouse
Immunogen Full length recombinant protein of human AlphaA Crystallin/CRYAA (NP_000385) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Membrane Trafficking and Chaperones, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1409.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.