Learn More
Description
Specifications
Specifications
| Antigen | AlphaB Crystallin/CRYAB |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.08 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:150. Use reported in scientific literature |
| Formulation | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide |
| Gene Accession No. | P02511 |
| Gene Alias | alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component |
| Gene Symbols | CRYAB |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
