Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaB Crystallin/CRYAB Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N102519A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102519A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102519A350 Supplier Novus Biologicals Supplier No. NB1002519AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AlphaB Crystallin/CRYAB Polyclonal specifically detects AlphaB Crystallin/CRYAB in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen AlphaB Crystallin/CRYAB
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Gene Symbols CRYAB
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human Crystallin AB protein (within residues 70-140). [Swiss-Prot P02511]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1410.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.