Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaB Crystallin/CRYAB Antibody (CPTC-CRYAB-1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB392376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392376 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392376 Supplier Novus Biologicals Supplier No. NBP308380FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen AlphaB Crystallin/CRYAB
Applications Western Blot, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone CPTC-CRYAB-1
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Host Species Mouse
Immunogen Recombinant human full-length AlphaB Crystallin/CRYAB protein (Uniprot: P02511)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1410
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.