Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaB Crystallin/CRYAB Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB1002519H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002519H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002519H Supplier Novus Biologicals Supplier No. NB1002519H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AlphaB Crystallin/CRYAB Polyclonal specifically detects AlphaB Crystallin/CRYAB in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen AlphaB Crystallin/CRYAB
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Accession No. P02511
Gene Alias alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Gene Symbols CRYAB
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human Crystallin AB protein (within residues 70-140). [Swiss-Prot P02511]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1410
Test Specificity Human and mouse. Predicted to react with rat, sheep, rabbit, monkey, guinea pig, chicken, and pig based on 100% sequence homology. Immunogen sequence has 81% homology with Zebrafish.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.